missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ninjurin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595755
This item is not returnable.
View return policy
Description
Immunogen sequence: MDSGTEEYEL NGGLPPGTPG SPDASPARWG WRHGPINVNH YASKKSAAES MLDIALLMAN ASQLKAVVEQ GPSFAFYVPL VVLISISLVL QIGVGVLLIF; Positive Samples: Mouse kidney, Rat spleen; Cellular Location: Membrane, Multi-pass membrane protein.
Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues. Cell adhesion requires divalent cations.
Specifications
| Ninjurin 1 | |
| Polyclonal | |
| Unconjugated | |
| Ninj1 | |
| AU024536; Nerve injury-induced protein 1; nerve injury-induced protein-1; NIN1; Ninj1; NINJURIN; ninjurin 1; ninjurin-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4814 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.959 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q92982 | |
| Ninj1 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction