missing translation for 'onlineSavingsMsg'
Learn More

Nintedanib esylate, MedChemExpress

Product Code. 16474398
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16474398

Brand: Medchem Express HY11106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nintedanib esylate (BIBF 1120 esylate) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively.

Nintedanib esylate (BIBF 1120 esylate) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively.

Specifications

Chemical Name or Material Nintedanib esylate
CAS 656247-18-6
Color Yellowish Green
Purity Grade Notes Research
Molecular Formula C33H39N5O7S
Quantity 10 mg
Synonym BIBF 1120 esylate
Solubility Information DMSO : 66.67 mg/mL (102.61 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (25.66 mM; Need ultrasonic) ∣Ethanol : 3.08 mg/mL (4.74 mM; Need ultrasonic)
SMILES O=C(C1=CC(NC/2=O)=C(C=C1)C2=C(NC3=CC=C(N(C)C(CN4CCN(C)CC4)=O)C=C3)/C5=CC=CC=C5)OC.CCS(=O)(O)=O
Molecular Weight (g/mol) 649.76
Formula Weight 649.76
Percent Purity 98.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.