missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NIP7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30477
This item is not returnable.
View return policy
Description
NIP7 Polyclonal antibody specifically detects NIP7 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| NIP7 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CGI-37,60S ribosome subunit biogenesis protein NIP7 homolog, HSPC031, KD93FLJ10296, nuclear import 7 homolog (S. cerevisiae) | |
| Between 130 and 180 | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51388 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction