missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NIPBL Antibody [FITC], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-93320F
This item is not returnable.
View return policy
Description
NIPBL Polyclonal antibody specifically detects NIPBL in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| NIPBL | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| FITC | |
| CDLS1, delangin, DKFZp434L1319, FLJ11203, FLJ12597, FLJ13354, FLJ13648, FLJ44854, IDN3-B, IDN3CDLS, Nipped-B homolog (Drosophila), nipped-B-like protein, Scc2, SCC2 homolog, sister chromatid cohesion 2 homolog | |
| The immunogen recognized by this antibody maps to a region between residue 1025 and 1075 of human Nipped-B-like using the numbering given in entry NP_597677.2 (GeneID 25836). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 25836 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction