Learn More
Invitrogen™ Nipocalimab Recombinant Monoclonal Antibody
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Nipocalimab works by binding to the neonatal Fc receptor (FcRn), inhibiting its interaction with immunoglobulin G (IgG), thereby reducing the levels of pathogenic IgG antibodies.
Specifications
Specifications
| Antigen | Nipocalimab |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 25mM histidine with 0.01% Tween 80, 8% sucrose and no preservative; pH 6.2 |
| Gene Alias | JNJ 80202135; M 281 |
| Host Species | Human |
| Purification Method | Protein A |
| Quantity | 1 mg |
| Show More |