Learn More
Ismatec™ Tubing Adaptor for Gear Pumpheads
Product Code. 10401241
Click to view available options
For Use With:
MCP Z Standard Gear Pumpheads
Unit Size:
Each
Product Code. 10401241
Brand: Ismatec™ AR0001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Connect the appropriate tubing to your gear pumphead with this adaptor. Ismatec™ Tubing Adaptor for Gear Pumpheads is constructed of threaded and barbed stainless steel.
  • Order in pairs to outfit your pumphead.
  • For MCP Z Standard Gear Pumpheads.
  • Threaded and barbed stainless steel construction.
Order Info
Type
Specifications
Description Thread size: 1/8in. NPT
For Use With MCP Z Standard Gear Pumpheads
Material Stainless Steel
Tubing Size 6 to 7 mm I.D.