Learn More
Ismatec™ Tubing Adaptor for Gear Pumpheads
Click to view available options
For Use With:
MCP Z Standard Gear Pumpheads
Unit Size:
Each
Description
- Order in pairs to outfit your pumphead.
- For MCP Z Standard Gear Pumpheads.
- Threaded and barbed stainless steel construction.
Order Info
Type
Specifications
Specifications
| Description | Thread size: 1/8in. NPT |
| For Use With | MCP Z Standard Gear Pumpheads |
| Material | Stainless Steel |
| Tubing Size | 6 to 7 mm I.D. |