missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIPSNAP1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76888FR
This item is not returnable.
View return policy
Description
NIPSNAP1 Polyclonal antibody specifically detects NIPSNAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NIPSNAP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| nipsnap homolog 1 (C. elegans), NIPSNAP, C. elegans, homolog 1,4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 1, NipSnap1, protein NipSnap homolog 1 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the center of human NIPSNAP. The immunogen is located within amino acids 130 - 180 of NIPSNAP. Amino Acid Squence: CMNKLKNNKEYLEFRRERT | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8508 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction