missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIPSNAP3A Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76968UV
This item is not returnable.
View return policy
Description
NIPSNAP3A Polyclonal antibody specifically detects NIPSNAP3A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NIPSNAP3A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| DKFZp564D177, FLJ13953, HSPC299, MGC14553, nipsnap homolog 3A (C. elegans), NipSnap3A, NipSnap4, protein NipSnap homolog 3A, Protein NipSnap homolog 4, Target for Salmonella secreted protein C, TassC | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human NIPSNAP3A. The immunogen is located within amino acids 40 - 90 of NIPSNAP3A. Amino Acid Squence: EFLENFEKNAHLRTAH | |
| 0.1 mL | |
| Neuroscience | |
| 25934 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction