missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIPSNAP3B Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-77341CL1
This item is not returnable.
View return policy
Description
NIPSNAP3B Polyclonal antibody specifically detects NIPSNAP3B in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| NIPSNAP3B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| FLJ11275, nipsnap homolog 3B (C. elegans), NIPSNAP3, NipSnap3B, protein NipSnap homolog 3B, SNAP1 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human NIPSNAP3B. The immunogen is located within amino acids 40 - 90 of NIPSNAP3B. Amino Acid Squence: AFMENLKKNIHLRTSY | |
| 0.1 mL | |
| Neuroscience | |
| 55335 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction