missing translation for 'onlineSavingsMsg'
Learn More

Niraparib, MedChemExpress

Product Code. 16426777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426777 5 mg 5mg
16465046 100 mg 100mg
16460427 50 mg 50mg
16404078 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16426777 Supplier Medchem Express Supplier No. HY10619

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Niraparib (MK-4827) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity.

Niraparib (MK-4827) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity.

Specifications

Chemical Name or Material Niraparib
CAS 1038915-60-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H20N4O
Quantity 5 mg
Synonym MK-4827
Solubility Information DMSO : 25 mg/mL (78.03 mM; Need ultrasonic)
SMILES NC(C1=CC=CC2=CN(C3=CC=C([C@H]4CNCCC4)C=C3)N=C21)=O
Molecular Weight (g/mol) 320.39
Formula Weight 320.39
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.