missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIRF Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93806-0.02ml
This item is not returnable.
View return policy
Description
NIRF Polyclonal antibody specifically detects NIRF in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NIRF | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| DKFZp434B0920, DKFZp686G0837, E3 ubiquitin-protein ligase UHRF2, EC 6.3.2, EC 6.3.2.-, MGC33463, NIRFUbiquitin-like-containing PHD and RING finger domains protein 2, Np95/ICBP90-like RING finger protein, Np95-like RING finger protein, Nuclear protein 97, Nuclear zinc finger protein Np97, RNF107RING finger protein 107, Ubiquitin-like PHD and RING finger domain-containing protein 2, ubiquitin-like with PHD and ring finger domains 2, ubiquitin-like, containing PHD and RING finger domains, 2, URF2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 76-359 of mouse NIRF (NP_659122.2). DSSLPSTSKQNDAQVKPSSHNPPKVKKTARGGSSSQPSTSARTCLIDPGFGLYKVNELVDARDVGLGAWFEAHIHSVTRASDGHSRGKTPLKNGSSYKRTNGNVNHNSKENTNKLDNVPSTSNSDSVAADEDVIYHIEYDEYPESGILEMNVKDLRPRARTILKWNELNVGDVVMVNYNVENPGKRGFWYDAEITTLKTISRTKKEVRVKVFLGGSEGTLNDCRVMSVDEIFKIEKPGAHPISFADGKFLRKNDPECDLCGGDPDKTCHMCSCHKCGEKRDPNM | |
| 0.02 mL | |
| Cancer, Zinc Finger | |
| 115426 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction