Learn More
NISCH Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | NISCH |
|---|---|
| Dilution | Western Blot 1:200 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NISCH Polyclonal antibody specifically detects NISCH in Human, Mouse samples. It is validated for Western BlotSpecifications
| NISCH | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 11188 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| hIRAS, I-1 receptor candidate protein, I-1FLJ14425, I1R, I1R candidate protein, Imidazoline receptor 1, imidazoline receptor antisera selected, Imidazoline receptor antisera-selected protein, imidazoline receptor candidate, Imidazoline-1 receptor, Imidazoline-1 receptor candidate protein, IR1, IRASFLJ40413, KIAA0975FLJ90519, nischarin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 500-700 of human NISCH (NP_009115.2). SLPQPILSNQGIMFVQEEALASSLSSTDSLTPEHQPIAQGCSDSLESIPAGQAASDDLRDVPGAVGGASPEHAEPEVQVVPGSGQIIFLPFTCIGYTATNQDFIQRLSTLIRQAIERQLPAWIEAANQREEGQGEQGEEEDEEEEEEEDVAENRYFEMGPPDVEEEEGGGQGEEEEEEEEDEEAEEERLALEWALGADEDF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |