Learn More
Invitrogen™ NIT1 Monoclonal Antibody (OTI4D12)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525068
Product Code. 15934674
  • € 572.00 / 100µL
Description
NIT1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Plays a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. It is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NIT1
Monoclonal
0.66 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q86X76
NIT1
Full length human recombinant protein of NIT1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI4D12
Unconjugated
NIT1
AI255805; Deaminated glutathione amidase; dGSH amidase; ESTM30; MGC57670; Nit1; nitrilase 1; nitrilase homolog 1; rCG_20134; W57327
Mouse
Affinity Chromatography
RUO
4817
-20°C, Avoid Freeze/Thaw Cycles