missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIT1 Monoclonal Antibody (OTI4D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501034
This item is not returnable.
View return policy
Description
Plays a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. It is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.Specifications
| NIT1 | |
| Monoclonal | |
| 0.66 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86X76 | |
| NIT1 | |
| Full length human recombit protein of human NIT1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4D12 | |
| Unconjugated | |
| NIT1 | |
| nitrilase 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4817 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction