missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIT2 Antibody (OTI2B9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72995CL1
This item is not returnable.
View return policy
Description
NIT2 Monoclonal antibody specifically detects NIT2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| NIT2 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.5.1.3, MGC111199, Nit protein 2, nitrilase family, member 2, Nitrilase homolog 2, omega-amidase NIT2 | |
| Full length human recombinant protein of human NIT2 (NP_064587) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 56954 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI2B9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction