Learn More
NIT2 Monoclonal Antibody (OTI3F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501098S
Encompass_Preferred
 View more versions of this product
Product Code. 17533291
  • € 242.00 / 30µL
Description
NIT2 Monoclonal antibody specifically detects NIT2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
NIT2
Monoclonal
0.72 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NQR4
Mouse
Affinity chromatography
RUO
56954
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI3F3
Unconjugated
Nit2
1190017B19Rik; CUA002; D16Ertd502e; epididymis secretory sperm binding protein Li 8a; HEL-S-8a; Nit protein 2; NIT2; nitrilase family member 2; nitrilase family, member 2; nitrilase homolog 2; Omega-amidase NIT2; RGD1310494
Full length human recombinant protein of human NIT2 produced in HEK293T cell.
30 μL