missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NIT2 Monoclonal Antibody (OTI5A3)
GREENER_CHOICE

Product Code. 15964694
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15964694 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15964694 Supplier Invitrogen™ Supplier No. MA525091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NIT2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Has a omega-amidase activity. The role of omega-amidase is to remove potentially toxic intermediates by converting alpha-ketoglutaramate and alpha-ketosuccinamate to biologically useful alpha-ketoglutarate and oxaloacetate, respectively. Overexpression decreases the colony-forming capacity of cultured cells by arresting cells in the G2 phase of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIT2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI5A3
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Nit2
Gene Accession No. Q9NQR4
Gene Alias 1190017B19Rik; CUA002; D16Ertd502e; epididymis secretory sperm binding protein Li 8a; HEL-S-8a; Nit protein 2; NIT2; nitrilase family member 2; nitrilase family, member 2; nitrilase homolog 2; Omega-amidase NIT2; RGD1310494
Gene Symbols Nit2
Host Species Mouse
Immunogen Full length human recombinant protein of NIT2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56954
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.