Learn More
Tocris Bioscience™ Nitazoxanide
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Nitazoxanide is a pyruvate flavodoxin/ferredoxin oxidoreductase (PFOR) inhibitor. Also inhibits Wnt signaling by stabilization of peptidyl arginine deiminase 2 (PAD2). Broad-spectrum antiparasitic and antiviral. Inhibits SARS-CoV-2 infection in Vero E6 cells (EC50 = 2.12μM).
Specifications
Specifications
| Chemical Name or Material | 2-(Acetyloxy)-N-(5-nitro-2-thiazolyl)benzamide |
| CAS | 55981-09-4 |
| Quantity | 50 mg |
| Target | Antiviral Compounds |
| Molecular Formula | C12H9N3O5S |
| Purity | 0.98 |