missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach™ NitraVer™ 5Nitrate Reagent Powder Pillows
Description
Pre-measured for 10mL samples.
Order Info
dimProductType
Specifications
Specifications
| Analytes | NO3-N |
| Format | Powder Pillow |
| For Use With (Application) | Determination of MR and HR nitrate |
| For Use With (Equipment) | All except DR/3000, DR/2010, DR/2000 |
| No. of Tests | 100 |
| Quantity | 100 Pack |
| Range | 0.3 - 30.0mg/L and 0.1 - 10.0mg/L |
| Sample Size | 10 mL |
| Storage Requirements | 10°C to 25°C |
| Test Method | 8039 HR, 8171 MR - Cadmium reduction |
| Show More |
Safety and Handling
Product Identifier
- NitraVer 5 Nitrate Reagent
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- H331-Toxic if inhaled.
- H410-Very toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P391-Collect spillage.
- MIXTURE LIST-Contain: Potassium dihydrogenphosphate, 4-aminobenzenesulphonic acid, sulphanilic acid,2, 5-Dihydroxybenzoi
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?