Learn More
WTW™ Nitrate Test Kits
Obtain precision measurements for waste water, drinking water, and surface water with CombiCheck Nitrate (NO3) cell test kits.
Brand: WTW™ 250444
This item is not returnable.
View return policy
Description
- Variety of instrumentation for measuring nitrates
- For waste water, drinking water, surface water
- PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, Photoflex photometers
- CombiCheck Nitrate (NO<sub>3</sub>) cell test kit
Specifications
Digestion with Persulfate | |
For PhotoLab™ S12, PhotoLab™ Spectral photometers | |
50 | |
50 Tests | |
Total Nitrogen NTotal Cuvette Test Kit |
For waste water, drinking water, surface water | |
Certificates | |
0.2 to 20 mg/L NO3-N, 0.9 to 88.5 mg/L NO3 | |
Photometric |