missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ NitriVer™ 3 Nitrite Reagent Powder Pillows

Product Code. 15364865
Change view
Click to view available options
Quantity:
100 Pack/100 Tests
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15364865 100 Pack/100 Tests Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15364865 Supplier Hach Lange™ Supplier No. 2107169
Outlet Corner
Use promo code 28080
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use, premeasured powder reagent in individual sealed packages for low range nitrite determination by the NitriVer™ 3 diazotization method. US EPA accepted for reporting wastewater analysis.

Ready-to-use, premeasured powder reagent in individual sealed packagesfor low range nitrite determination by the NitriVer™ 3 diazotization method.US EPA accepted for reporting wastewater analysis.

Powder Pillows make titration applications quick and easy.

  • Sealed packets ensure fresh, uncontaminated reagent every time
  • Premeasured to avoid measurement error and contamination from handling
  • Ready to use packets save time and hassle
  • Exact doses to easily monitor supply and prepare for lab procedures
  • Single-use packs for easy transportation and packing in test kits
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
For Use With (Equipment) All Hach colorimeters except PC II
Packaging Individual Packets
Quantity 100 Pack/100 Tests
Test Method Diazotization
compliance-icons compliance-icons
Product Identifier
  • NitriVer 3 Nitrite Reagent
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: potassium dihydrogenphosphate, potassium disulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.