missing translation for 'onlineSavingsMsg'
Learn More

Nitrotryptophan Antibody (2D12), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18477844 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18477844 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18477844 Supplier Novus Biologicals Supplier No. NBP259354MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nitrotryptophan Monoclonal antibody specifically detects Nitrotryptophan in Non-species specific samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nitrotryptophan
Applications Flow Cytometry
Classification Monoclonal
Clone 2D12
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias 6-Nitrotryptophan;6-NO2-Trp;Nitrotryptophan;NW
Host Species Mouse
Immunogen Synthetic 6-Nitrotryptophan modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.