missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrotyrosine Antibody (39B6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-96877B
This item is not returnable.
View return policy
Description
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Rat, Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Nitrotyrosine | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Rat, Non-species specific | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 39B6 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Nitrotyrosine | |
| Hybridoma line 39B6 | |
| 0.1 mL | |
| Recognizes 3-nitrotyrosine moieties. No detectable cross-reactivity with non-nitrated tyrosine. Not species specific. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction