missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrotyrosine Antibody (39B6), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-96877MFV500
This item is not returnable.
View return policy
Description
Nitrotyrosine Monoclonal antibody specifically detects Nitrotyrosine in Rat, Non-species specific samples. It is validated for Flow Cytometry
Specifications
| Nitrotyrosine | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Rat, Non-species specific | |
| Purified |
| Flow Cytometry | |
| 39B6 | |
| Flow Cytometry | |
| Nitrotyrosine | |
| 3-(4-hydroxy-3-nitrophenylacetamido) propionic acid-bovine serum albumin | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction