Learn More
Nitrotyrosine Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB3248-SP
Product Code. 18786498
  • € 210.00 / 25µg
Description
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Multi-Species samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nitrotyrosine
Monoclonal
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Nitrotyrosine
Nitrotyrosine-modified KLH
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG3
Western Blot, Immunocytochemistry
306507
Unconjugated
Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Detects Nitrotyrosine adducts on proteins in Western blots. It does not cross-react with phosphotyrosine or 4-hydroxynonenal adducts. Unfixed cells, tissues, and proteins can be treated with Peroxynitrite (Catalog # AR006 ) for use as positive controls with this antibody.
Multi-species