missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Njmu-R1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521978
This item is not returnable.
View return policy
Description
Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
As component of the WDR11 complex acts together with TBC1D23 to facilitate the golgin-mediated capture of vesicles generated using AP-1 (PubMed:29426865). May have a role in spermatogenesis. [UniProt].
Specifications
| Njmu-R1 | |
| Polyclonal | |
| Unconjugated | |
| C17orf75 | |
| 5730455P16Rik; C17orf75; chromosome 17 open reading frame 75; NJMU-R1; Protein Njmu-R1; RGD1310429; RIKEN cDNA 5730455P16 gene; sensitization to ricin complex subunit 2; similar to Protein Njmu-R1; spermatogenesis-related protein; SRI2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64149, 70591 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9CYI0, Q9HAS0 | |
| 5730455P16Rik, C17orf75 | |
| Recombinant protein encompassing a sequence within the center region of human C17orf75. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction