Learn More
NK1R Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB300-119PE
Product Code. 18740194
  • ONLINE EXCLUSIVE PRICE
    € 680.00
    € 642.60 / 0.10mL
    Save € 37.40
    5% Off
Description
NK1R Polyclonal specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NK1R
Polyclonal
Flow Cytometry
P25103
TACR1
A peptide derived from the N-terminus region between amino acids 1-50 of human Neurokinin 1 Receptor. [UniProt# P25103]
Affinity Purified
RUO
Primary
Reacts with human, mouse and rat. Immunogen has 94% sequence identity with guinea pig. Mouse reactivity is from a customer review.
Store at 4C in the dark.
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor
Rabbit
46 kDa
0.1 mL
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
6869
Human, Mouse, Rat