Learn More
NK2R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69018
 View more versions of this product
Product Code. 18630868
  • € 589.00 / 100µL
Description
NK2R Polyclonal antibody specifically detects NK2R in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NK2R
Polyclonal
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Neurokinin A receptor, NK2RNK-2 receptor, NKNARseven transmembrane helix receptor, SKRneurokinin 2 receptor, substance-K receptor, TAC2Rsubstance K receptor, tachykinin receptor 2NK-2R
This antibody was developed against a recombinant protein corresponding to amino acids: STVTMDQGATKCVVAWPEDSGGKTLLL
100 μg
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
6865
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Protein A purified