missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK3R/TACR3/Neurokinin B Receptor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 231.00 - € 345.00
Specifications
| Antigen | NK3R/TACR3/Neurokinin B Receptor |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600588
|
Novus Biologicals
NBP3-06055-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680499
|
Novus Biologicals
NBP3-06055-200ul |
200 μg |
€ 345.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694059
|
Novus Biologicals
NBP3-06055-50ul |
50 μg |
€ 231.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NK3R/TACR3/Neurokinin B Receptor Polyclonal antibody specifically detects NK3R/TACR3/Neurokinin B Receptor in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| NK3R/TACR3/Neurokinin B Receptor | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS, pH7.0 | |
| 6870 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Neurokinin B receptor, neurokinin beta receptor, neuromedin-K receptor, NK-3 receptor, NK-3R, NK3RMGC148061, NKR, TAC3R, TAC3RL, tachykinin receptor 3MGC148060 | |
| Produced in rabbits immunized with E. coli-derived Human NK3R/TACR3/Neurokinin B Receptor fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title