missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKCC1/SLC12A2 Antibody (915911) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8528F
This item is not returnable.
View return policy
Description
NKCC1/SLC12A2 Monoclonal antibody specifically detects NKCC1/SLC12A2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| NKCC1/SLC12A2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6558 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 915911 | |
| Flow Cytometry, CyTOF-ready | |
| Basolateral Na-K-Cl symporter, BSC, BSC2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, NKCC1MGC104233, solute carrier family 12 (sodium/potassium/chloride transporters), member 2, solute carrier family 12 member 2 | |
| NS0 mouse myeloma cell line transfected with human NKCC1/SLC12A2, Met1-Ser1212, Accession # P55011 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction