missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NKCC2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142445
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
The sodium-potassium-chloride cotransporter isoform 2 is kidney-specific and is found on the apical membrane of the thick ascending limb of Henle's loop and the macula densa. It accounts for most of the NaCl resorption with the stoichiometry of 1Na:1K:2Cl and is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene.
Specifications
| NKCC2 | |
| Polyclonal | |
| Unconjugated | |
| SLC12A1 | |
| AI788571; apical Na(2Cl)K cotransporter; BSC1; bumetanide-sensitive cotransporter type 1; bumetanide-sensitive Na-K-Cl cotransport protein splice isoform A; bumetanide-sensitive Na-K-Cl cotransport protein splice isoform B; bumetanide-sensitive Na-K-Cl cotransport protein splice isoform F; bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; D630042G03Rik; Kidney-specific Na-K-Cl symporter; mBSC1; MGC48843; Na-K-2Cl cotransporter; NKCC2; NKCC2A variant A; SLC12A1; solute carrier family 12 (sodium/potassium/chloride transporter), member 1; solute carrier family 12 (sodium/potassium/chloride transporters), member 1; solute carrier family 12 member 1; Solute carrier family 12 member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter); solute carrier family 12, member 1; Solute carrier family 12, member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter); urehr3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 100154291, 20495, 6557 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P55014, Q13621 | |
| SLC12A1 | |
| Peptide with sequence C-PHYEETSFGDEAQKR. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction