missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKCC2/SLC12A1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98480FR
This item is not returnable.
View return policy
Description
NKCC2/SLC12A1 Polyclonal antibody specifically detects NKCC2/SLC12A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NKCC2/SLC12A1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| BSC1, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2, Kidney-specific Na-K-Cl symporter, MGC48843, NKCC2A variant A, NKCC2Na-K-2Cl cotransporter, solute carrier family 12 (sodium/potassium/chloride transporters), member 1, solute carrier family 12 member 1 | |
| Produced in rabbits immunized with E. coli-derived Human NKCC2/SLC12A1 fragment. | |
| 0.1 mL | |
| ABC Transporters, Cancer, Neuroscience | |
| 6557 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction