Learn More
NKG2A/C/E Monoclonal Antibody (20d5), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5896-025
Description
Description: The 20d5 monoclonal antibody reacts with the mouse NKG2A, C, and E. NKG2 molecules belong to a C-type lectin-like family of cell surface receptors expressed by mouse NK and NKT cell lineages. NKG2 molecules form heterodimeric complexes with CD94 and are responsible for recognition of non-classical MHC class I antigen Qa-1. When co-staining with NKG2A^B6 clone 16a11, it is important to stain for NKG2A-B6 first then subsequently with NKG2A/C/E, as steric hindrance has been observed if co-stained concurrently. Applications Reported: This 20d5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 20d5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
NKG2A/C/E Monoclonal antibody specifically detects NKG2A/C/E in Mouse samples. It is validated for Control, Flow Cytometry, Functional Assay
Specifications
| NKG2A/C/E | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| CD159a; cd159c; cd159e; Cluster of Differentiation 159a; killer cell lectin-like receptor subfamily C, member 1; killer cell lectin-like receptor subfamily C, member 2; killer cell lectin-like receptor subfamily C, member 3; Klrc1; Klrc2; Klrc3; natural killer cell group 2E cell receptor; NKG2; NKG2A; NKG2-A/NKG2-B type II integral membrane protein; NKG2B; NKG2C; Nkg2e | |
| Affinity chromatography | |
| RUO | |
| 16641, 16642, 58179 | |
| 4° C | |
| Liquid |
| Flow Cytometry, Functional Assay | |
| 20d5 | |
| Functional Grade | |
| Klrc3 | |
| Rat | |
| 25 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.