missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKG2A/CD159a/KLRC1 Antibody (131411R) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1059RC
This item is not returnable.
View return policy
Description
NKG2A/CD159a/KLRC1 Monoclonal antibody specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| NKG2A/CD159a/KLRC1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 131411R | |
| Flow Cytometry, CyTOF-ready | |
| CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
| BaF3 mouse pro-B cell line transfected with human NKG2A and CD94 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| 3821 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction