Learn More
NKG2A (CD159a) Mouse anti-Human, Brilliant Violet 510, Clone: 131411, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747922
This item is not returnable.
View return policy
Description
NKG2A (CD159a) Monoclonal antibody specifically detects NKG2A (CD159a) in Human samples. It is validated for Flow Cytometry.
Specifications
| NKG2A (CD159a) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| NKG2A (CD159a) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 131411 | |
| Brilliant Violet 510 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG2a κ |