Learn More
NKG2A (CD159a) Mouse anti-Human, Brilliant Violet 510, Clone: 131411, BD OptiBuild™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 747922
Product Code. 17259564
  • € 530.00 / 50µg
Description
NKG2A (CD159a) Monoclonal antibody specifically detects NKG2A (CD159a) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKG2A (CD159a)
Monoclonal
0.2 mg/mL
Mouse
RUO
NKG2A (CD159a)
Refrigerate (2 to 8°C)
Flow Cytometry
131411
Brilliant Violet 510
50 μg
Primary
Human
IgG2a κ