Learn More
NKG2D/CD314 Antibody (6E5A7) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20156-0.2mg
 View more versions of this product
Product Code. 18679803
  • € 635.00 / 0.20mg
Description
NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKG2D/CD314
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, Immunofluorescence, Neutralization
6E5A7
Unconjugated
CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence
The original antibody was generated by immunizing BALB/c mice with soluble NKG2-D fragments containing amino acids from 64-462 and 123-462.
0.2 mg
Adaptive Immunity, Immunology, Neuroscience, Signal Transduction, Stem Cells
22914
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ