missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKIAMRE Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-123V
This item is not returnable.
View return policy
Description
NKIAMRE Polyclonal antibody specifically detects NKIAMRE in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| NKIAMRE | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DyLight 405 | |
| cyclin-dependent kinase-like 3, EC 2.7.11, EC 2.7.11.22, NKIAMRESerine/threonine-protein kinase NKIAMRE, serine-threonine protein kinase NKIAMRE | |
| Synthetic peptide corresponding to residues 414-428 [NCNGLKENPHCGGSV] of the 50 kDa human NKIAMRE protein. | |
| 0.1 mL | |
| Cancer, Protein Kinase, Tumor Suppressors | |
| 51265 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction