missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKIRAS1 Antibody (184C278), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56555AF750
This item is not returnable.
View return policy
Description
NKIRAS1 Monoclonal antibody specifically detects NKIRAS1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Primate, Xenopus samples. It is validated for Western Blot
Specifications
| NKIRAS1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| Western Blot | |
| 184C278 | |
| Western Blot | |
| I-kappa-B-interacting Ras-like protein 1, kappa B-ras 1, Kappa B-Ras protein 1, KappaB-Ras1, KBRAS1kappaB-Ras1, NF-kappa-B inhibitor-interacting Ras-like protein 1, NFKB inhibitor interacting Ras-like 1, NFKB inhibitor interacting Ras-like protein 1 | |
| This monoclonal antibody was raised against a synthetic peptide containing amino acid residues 96-113 (RVELLKKEIDKFKDKKEV) of human kB-Ras1. | |
| 0.1 mL | |
| Signal Transduction | |
| 28512 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction