missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp30/NCR3 Antibody (2615A), PE/Cy7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB18492PECY7
This item is not returnable.
View return policy
Description
NKp30/NCR3 Monoclonal antibody specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry
Specifications
| NKp30/NCR3 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2615A | |
| Flow Cytometry | |
| Activating natural killer receptor p30, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NKP30, NKp30MALS | |
| Mouse myeloma cell line NS0-derived human NKp30/NCR3, Leu19-Thr138, Accession # Q05D23 | |
| 0.1 mL | |
| Adaptive Immunity, Checkpoint signaling, Immunology, Stem Cells | |
| 259197 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction