Learn More
NKp30/NCR3 Antibody (MM0490-9D31), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-11819H
Product Code. 18475092
  • € 607.00 / 0.10mL
Description
NKp30/NCR3 Monoclonal antibody specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HRP
PBS
259197