missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp44/NCR2 Antibody (253415), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB22491F
This item is not returnable.
View return policy
Description
NKp44/NCR2 Monoclonal antibody specifically detects NKp44/NCR2 in Human samples. It is validated for Flow Cytometry, Agonist Activity, CyTOF-reported
Specifications
| NKp44/NCR2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Agonist Activity, CyTOF | |
| 253415 | |
| Flow Cytometry, Agonist Activity, CyTOF-reported | |
| CD336, CD336 antigen, LY95lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44), lymphocyte antigen 95 (activating NK-receptor; NK-p44), Lymphocyte antigen 95 homolog, natural cytotoxicity triggering receptor 2, Natural killer cell p44-related protein, NK cell activating receptor (NKp44), NK cell-activating receptor, NKP44, NK-p44dJ149M18.1 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp44/NCR2, Gln22-Pro190, Accession # CAB52289 | |
| 0.1 mL | |
| Innate Immunity | |
| 9436 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction