missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp44/NCR2 Antibody (SR1752), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | NKp44/NCR2 |
|---|---|
| Clone | SR1752 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694284
|
Novus Biologicals
NBP3-22479-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681945
|
Novus Biologicals
NBP3-22479-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKp44/NCR2 Monoclonal antibody specifically detects NKp44/NCR2 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| NKp44/NCR2 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD336, CD336 antigen, LY95lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44), lymphocyte antigen 95 (activating NK-receptor; NK-p44), Lymphocyte antigen 95 homolog, natural cytotoxicity triggering receptor 2, Natural killer cell p44-related protein, NK cell activating receptor (NKp44), NK cell-activating receptor, NKP44, NK-p44dJ149M18.1 | |
| A synthesized peptide derived from human NKp44/NCR2 (Uniprot #: O95944) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1752 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Innate Immunity | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9436 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title