Learn More
NKp46/NCR1 Antibody (195314), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1850MFV450
Product Code. 18509888
  • € 695.00 / 0.10mL
Description
NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Agonist Activity, Immunocytochemistry, CyTOF-reported
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKp46/NCR1
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
0.1 mL
Adaptive Immunity, Immunology, Signal Transduction
9437
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Agonist Activity, Immunocytochemistry
195314
Western Blot, Flow Cytometry, Immunohistochemistry, Agonist Activity, Immunocytochemistry, CyTOF-reported
CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Mouse T cell hybridoma transfected with human NKp46/NCR1
RUO
Primary
Human
IgG2b