missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp46/NCR1 Antibody (29A1.4), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB22252K
This item is not returnable.
View return policy
Description
NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-reported
Specifications
| NKp46/NCR1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 29A1.4 | |
| Flow Cytometry, CyTOF-reported | |
| CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| Murine myeloma cell line NSO-derived recombinant mouse NKp46-Fc chimera , Glu22-Asn255, Accession # Q8C567 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| 9437 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction