missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp46/NCR1 Antibody (B-L46), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14597JF646
This item is not returnable.
View return policy
Description
NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry
Specifications
| NKp46/NCR1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| Recombinant human NKp46/NCR1 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| 9437 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| B-L46 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction