Learn More
NKp80/KLRF1 Antibody (239127) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB1900JF635
Product Code. 30171073
  • € 837.00 / 0.50mL
Description
NKp80/KLRF1 Monoclonal antibody specifically detects NKp80/KLRF1 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKp80/KLRF1
Monoclonal
Janelia Fluor 635
Activating coreceptor NKp80, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80
Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1, Val66-Tyr231, Accession # Q9NZS2
0.5 mL
Immunology
51348
Store at 4°C in the dark.
IgG2a
Flow Cytometry, CyTOF
239127
50mM Sodium Borate
Rat
Protein A or G purified