missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp80/KLRF1 Antibody (239127) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1900JF669
This item is not returnable.
View return policy
Description
NKp80/KLRF1 Monoclonal antibody specifically detects NKp80/KLRF1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| NKp80/KLRF1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Activating coreceptor NKp80, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1, Val66-Tyr231, Accession # Q9NZS2 | |
| 0.5 mL | |
| Immunology | |
| 51348 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 239127 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction