missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp80/KLRF1 Antibody (MM0492-7D32), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11821C
This item is not returnable.
View return policy
Description
NKp80/KLRF1 Monoclonal antibody specifically detects NKp80/KLRF1 in Human samples. It is validated for Flow Cytometry.
Specifications
| NKp80/KLRF1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KLRF1 | |
| Human recombinant NKp80/KLRF1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| MM0492-7D32 | |
| Flow Cytometry | |
| Activating coreceptor NKp80, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80 | |
| Rat | |
| Protein G purified | |
| Immunology | |
| 51348 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction