Learn More
NKR-P1 Antibody (3.2.3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22437-100ug
This item is not returnable.
View return policy
Description
NKR-P1 Monoclonal specifically detects NKR-P1 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| NKR-P1 | |
| Monoclonal | |
| 0.51 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready | |
| KLRB1 | |
| Rat 24.5 kD cell surface protein known as NKR-P1 | |
| 100 ug | |
| Primary | |
| This antibody reacts with Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| 3.2.3 | |
| Unconjugated | |
| killer cell lectin-like receptor subfamily B, member 1, Klrb1, Klrb1 killer cell lectin-like receptor subfamily B, member 1, Klrb1d, Klrb1f, Klrb1g, Klrb6, Nkr-p1e, Nkrp-1e, NKR-P1G | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 689817 | |
| Rat | |
| Purified |