missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKRF Antibody (OTI4C2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73001
This item is not returnable.
View return policy
Description
NKRF Monoclonal specifically detects NKRF in Human, Mouse samples. It is validated for Western Blot.
Specifications
| NKRF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| ITBA4NFkB-repressing factor, NF-kappaB repressing factor, NFKB repressing factor, NRFNF-kappa-B-repressing factor, Protein ITBA4, Transcription factor NRF | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot | |
| OTI4C2 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| NKRF | |
| Human recombinant protein fragment corresponding to amino acids 1-296 of human NKRF(NP_060014) produced in E.coli. | |
| 100 μg | |
| 55922 | |
| Human, Mouse | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction